Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RUFY2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113256
Description
RUFY2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
Specifications
RUFY2 | |
Polyclonal | |
Unconjugated | |
RUFY2 | |
2610111M19Rik; AI844453; antigen MU-RMS-40.17; Denn; Kiaa1537; Leucine zipper FYVE-finger protein; LZ-FYVE; Rab4-interacting protein related; RABIP4R; RUFY2; RUN and FYVE domain containing 2; RUN and FYVE domain-containing 2; RUN and FYVE domain-containing protein 2; Run- and FYVE-domain containing protein; ZFYVE13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55680, 70432 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R4C2, Q8WXA3 | |
RUFY2 | |
Recombinant Human RUN and FYVE domain-containing protein 2 protein (396-655AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction