Learn More
RUFY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 191801AP
Catalog No. 50-173-2361
Add to cart
Description
Description
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
Specifications
Specifications
RUFY2
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R4C2, Q8WXA3
KIAA1537, RABIP4R, RUFY2, ZFYVE13
Rabbit
IgG
150 μL
-20°C
Primary
55680, 690777, 70432
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUFY2
Liquid
RUFY2
RUFY2 Fusion Protein Ag5938
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat