Learn More
Ruled Microscope Slides Products for Science Education
Keep tabs on your microorganisms.
Manufacturer:  Neo/Sci Corporation 704002
Catalog No. S66585
  Check Availability
Add to cart
Specifications
Description
  • Gridwork slide allows students to easily keep track of the contents
  • Count populations or follow the movements of particular specimens
  • Specially lined slides feature 64 2mm squares
  • Set of five slides
Description & Specifications
Specifications
Ruled Microscope Slide