Learn More
Cole-Parmer Parker Hannifin Fitting, PFA, Tee, Compression Branch to Threaded Adapter, 3/4" OD x 3/4" OD x 3/4" NPT(M)  DFS Item
Manufacturer:  Cole-Parmer 3132039
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal. These branch tees feature a male NPT thread on the bottom branch for quick connection to a threaded port.
Catalog No. 50-631-953