Learn More
Cole-Parmer Parker Hannifin GAMT-424 Compression Run Tee, PFA, 1/4" OD X 1/8" NPT(M)  DFS Item
Manufacturer:  Cole-Parmer 3132174
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal. Choose PFA material for enhanced chemical resistance. Black polypropylene is an economical option that can be autoclaved.
Catalog No. 50-631-956