Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expedeon RunBlue™ Bis-Tris Protein Gels 8%

For use with polyacrylamide gel electrophoresis of proteins based on Bis-Tris neutral gel buffer system.

$149.56 - $152.10

Specifications

Gel Type MES, MOPS
Gel Percentage 8%
Storage Requirements +4°C
For Use With (Application) Electrophoresis- Acrylamide or Bis Acrylamide
Length (Metric) Gel 10 cm
View More Specs

Products 2
Catalog Number Mfr. No. Wells Price Quantity & Availability  
Catalog Number Mfr. No. Wells Price Quantity & Availability  
NBT00812 View Documents Expedeon
NBT00812
12-well Pack of 10 for $152.10
Add to cart
 
NBT00827 View Documents Expedeon
NBT00827
17-well Pack of 10 for $149.56
Add to cart
 
Description

Description

  • Compatible with MES and MOPS buffers
  • Protein Gels have an approximate range of 3.5 kDa to 160 kDa with MES Buffer and 30 kDa to 180 kDa
  • Compatible with tanks such as NuPAGE™ XCell SureLock™ Mini-Cell
  • Fast run times at 200V
  • Sharp High resolution bands showing protein integrity and separation
  • Heterogeneous polymerisation across the gel due to acrylamide concentration with consistency and no residual free acrylamide
  • Samples up to 12 or 17 can be run simultaneously
  • No well contamination in case of overloading due to protruted teeth
  • Resilient tear-proof gel matrix ensures risk-free handling
  • Comb free feature ensures reduced risk of broken or damaged teeth
  • Strip free feature enusres no potentially ruined Western Blots
  • Shelf life of 1 year at 4°C and 3 months at Room Temperature
Specifications

Specifications

MES, MOPS
+4°C
10 cm
Bis-Tris Protein Gels
8%
Electrophoresis- Acrylamide or Bis Acrylamide
10 cm
Safety and Handling

Safety and Handling

ShelfLife : 1 year at 4°C and 3 months at Room Temperature

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit