Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNDC3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205311AP

Catalog No. 50-173-2941


Add to cart

Description

Description

May act as an effector of RAP2A in neuronal cells.
Specifications

Specifications

RUNDC3A
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08576, Q59EK9
Rap2 interacting protein 8, RAP2IP, RPIP 8, RPIP8, RUN domain containing 3A, RUNDC3A
Rabbit
IgG
150 μL
-20°C
Primary
10900, 303569, 51799
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUNDC3A
Liquid
Rundc3a
RUNDC3A Fusion Protein Ag14393
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit