Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RUNDC3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563407
Description
Immunogen sequence: EKSYQSLDQL SAEVSLSQTS LDPGQSQEGD GKQDTLNVMS EGKEDTPS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.
RUNDC3B is a protein coding gene.
Specifications
RUNDC3B | |
Polyclonal | |
Unconjugated | |
Rundc3b | |
Gm440; Rap2 binding protein 9; Rap2-binding protein 9; Rap2-interacting protein 9; RPIB9; RPIP9; RPIP-9; RUN domain containing 3B; RUN domain-containing protein 3B; rundc3b; zgc:101561 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
154661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96NL0 | |
Rundc3b | |
Recombinant protein corresponding to Human RUNDC3B. Recombinant protein control fragment (Product #RP-101731). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction