Learn More
Aviva Systems Biology Corp RUNX1T1 Antibody - middle region (ARP58127_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RUNX1T1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RUNX1T1. Alternate names: AML1T1, CBFA2T1, CDR, ETO, MGC2796, MTG8, MTG8b, ZMYND2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62244