Learn More
RUNX1T1/ETO Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259232
Catalog No. NBP259232
Add to cart
Description
Description
RUNX1T1/ETO Polyclonal antibody specifically detects RUNX1T1/ETO in Human samples. It is validated for Chromatin Immunoprecipitation, ELISA.
Specifications
Specifications
RUNX1T1/ETO
Unconjugated
Serum with 0.05% Sodium Azide
AML1T1protein CBFA2T1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, Protein ETO, Protein MTG8, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22
Rabbit
IgG
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
ChIP assay, ELISA
Chromatin Immunoprecipitation 4 ul, ELISA 1:100
Antiserum
RUNX1T1
The exact immunogen is propietary information.
Whole Antiserum
RUO
Primary
862