Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1T1/ETO Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen RUNX1T1/ETO
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123642
View Documents
Novus Biologicals
NBP30937025UL
25 μg
Each for $106.19
Add to cart
 
NB123641
View Documents
Novus Biologicals
NBP309370100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

RUNX1T1/ETO Polyclonal antibody specifically detects RUNX1T1/ETO in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

RUNX1T1/ETO
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
PBS buffer, 2% sucrose
862
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Human
AML1T1protein CBFA2T1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, Protein ETO, Protein MTG8, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22
The immunogen is a synthetic peptide directed towards the middle region of human RUNX1T1/ETO (NP_004340). Peptide sequence RQCNLQQFIQQTGAALPPPPRPDRGPPGTQGPLPPAREESLLGAPSESHA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit