Learn More
RUNX3/CBFA3 Mouse anti-Human, Alexa Fluor™ 594, Clone: 2B9G4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222390AF594
Catalog No. NP222390A94
Add to cart
Description
Description
RUNX3/CBFA3 Monoclonal antibody specifically detects RUNX3/CBFA3 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
RUNX3/CBFA3
2B9G4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Acute myeloid leukemia 2 protein, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3FLJ34510, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
RUNX3
Amino acids 126-310 of human Runx3 were used as the immunogen for this antibody.
0.1 mL
Primary
864