Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUSC2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548330

Catalog No. PA548330


Only null left
Add to Cart

Description

Description

RUSC2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a RUN and SH3 domain containing protein that interacts with Rab1b and Rab1-binding protein GM130. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RUSC2
Polyclonal
Unconjugated
RUSC2
AI840675; Interacting protein of Rab1; iporin; KIAA0375; mKIAA0375; RUN and SH3 domain containing 2; RUN and SH3 domain-containing protein 2; RUSC2
Rabbit
Antigen affinity chromatography, Protein A
RUO
9853
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q8N2Y8
RUSC2
KLH conjugated synthetic peptide between 308-336 amino acids from the N-terminal region of human RUSC2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.