Rusch Murphy™ Endotracheal Tube Airways
Manufacturer: Moore Medical Corp 15191
 View more versions of this product
Catalog No. 19-898-054
  Check Availability
Add to Cart
    Specifications
    Color Clear
    Connections Luer and Luer lock syringe tip and atraumatic tip
    Graduated Yes
    Includes Cuffs
    Material PVC
    Size 6mm
    Sterile Yes
    Product Type Airways Tube
  • Description & Specifications
Related Products
Product Content Feedback