Learn More
Sklar InstrumentsSupplier Diversity Partner Russian Tissue Forceps, Sklar, 10 in., Straight, Radially Serrated, Stainless Steel. Sklar Russian Forceps are medium-heavy forceps used for grasping heavy or thick tissue. They are also used during wound closure. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 192310
Russian Tissue Forceps, Sklar, 10 in., Straight, Radially Serrated, Stainless Steel. Sklar Russian Forceps are medium-heavy forceps used for grasping heavy or thick tissue. They are also used during wound closure. This product is straight with atraumatic, radially serrated, cupped tips. The length is 10 inches.
Catalog No. 50-143-7431