Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ruthenium, 0.5% on granular carbon
Click to view available options
Quantity:
25g
100g
Specifications
Concentration or Composition (by Analyte or Components) | 0.5% on granular carbon |
UN Number | UN1325 |
Solubility Information | Soluble in fused alkalis. Insoluble in water,acid and aqua regia. |
Physical Form | 4-12 Mesh,900-1100m |
Concentration | 0.5% on granular carbon |
Mesh Size | 4 to 12 Mesh |
Odor | Odorless |
Surface Area | 900 to 1100m2/g |
Quantity | 25g |
Chemical Name or Material | Ruthenium |
Safety and Handling
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Warning
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 231-127-1
TSCA : Yes
Recommended Storage : Ambient temperatures