Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous, ACROS Organics™

Click to view available options
Quantity:
2g
10g
50g
Packaging:
Glass bottle
10049-08-8
Cl3Ru
207.43
10049-08-8
Cl3Ru
207.43
for ultra trace analysis
Assay Percent Range 48 to 50% (Ru)
Linear Formula RuCl3
Fieser 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292
Merck Index 15,8436
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water,slightly soluble in alcohol
Formula Weight 207.43
Physical Form Powder or Crystals
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: decomposes in hot water, slightly soluble in alcohol
Total Trace Metal Impurities 0.1% max
Color Black
Melting Point >500.0°C
Quantity 50g
Chemical Name or Material Ruthenium(III) chloride
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 233-167-5