Alfa Aesar™ Ruthenium(III) chloride, anhydrous, Ru 47.7% min

Manufacturer: Alfa Aesar™ 4718209

Quantity 10g

 View more versions of this product

Catalog No. AA4718209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10049-08-8
    Linear Formula RuCl3
    Formula Weight 207.43
    MDL Number MFCD00011208
    UN Number UN3260
    Merck Index 14,8302
    Chemical Name or Material Ruthenium(III) chloride, Anhydrous

    Specifications

    Formula Weight 207.43
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011208
    TSCA TSCA
    Odor Pungent
    Physical Form Powder
    Sensitivity Hygroscopic
    CAS 10049-08-8
    Color Black
    Melting Point >500°C (dec.)
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    RTECS Number VM2650000
    UN Number UN3260
    Merck Index 14,8302
    Quantity 10g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H290-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    Concentration Ru 47.7% min
    Density 3.110g/mL
    Linear Formula RuCl3
    Chemical Name or Material Ruthenium(III) chloride, Anhydrous

    GHS H Statement
    H314-H318-H290-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit