Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) nitrosylnitrate, Ru 31.3% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1217506

Quantity 5g

 View more versions of this product

Catalog No. AA1217506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

34513-98-9
Ru 31.3% min.
317.09
Ruthenium(III) nitrosylnitrate
Crystalline
N4O10Ru
MFCD00016313

Specifications

34513-98-9
Ru 31.3% min.
Red-brown crystals
MFCD00016313
Red-brown
Ruthenium(III) nitrosylnitrate
Crystalline
N4O10Ru
317.09
Odorless
5g

Safety and Handling

SDS & Safety Documents

H272-H314-H318-H290: May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 252-068-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit