Alfa Aesar™ Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v

Manufacturer: Alfa Aesar™ 12630AU

Quantity 1000mL

 View more versions of this product

Catalog No. AA12630AU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34513-98-9
    Linear Formula Ru(NO)(NO3)3
    Formula Weight 317.09
    MDL Number MFCD00016313
    UN Number UN3093
    Chemical Name or Material Ruthenium(III) nitrosylnitrate solution

    Specifications

    Formula Weight 317.09
    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    Density 1.07g/mL
    MDL Number MFCD00016313
    CAS 34513-98-9
    UN Number UN3093
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H314
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
    Color Red-brown
    Quantity 1000mL
    Odor Acidic
    Concentration Ru 1.5% w/v
    Linear Formula Ru(NO)(NO3)3
    Chemical Name or Material Ruthenium(III) nitrosylnitrate solution

    GHS H Statement
    H272-H314
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit