Ruthenium(IV) oxide, Electronic Grade, 99.95% (Metals basis), Alfa Aesar™, Premion™

For Semiconductor Industry Use For the Manufacture of Resistor Pastes

Manufacturer: Alfa Aesar 4033602

Quantity 0.5g

 View more versions of this product

Catalog No. AA4033602

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-10-1
    Physical Form Powder
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula RuO2
    Formula Weight 133.07
    MDL Number MFCD00011210
    Synonym Ruthenium dioxide
    Chemical Name or Material Ruthenium(IV) oxide
    Grade Electronic Grade

    Specifications

    Physical Form Powder
    Formula Weight 133.07
    Quantity 0.5g
    Color Black
    DOT Information DOT Class: 5.1, Packaging Group: II
    EINECS Number 234-840-6
    Odor Odorless
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula RuO2
    Appearance Black powder
    MDL Number MFCD00011210
    CAS 12036-10-1
    Synonym Ruthenium dioxide
    Health Hazard 1 H271-H303: May cause fire or explosion; strong oxidizer. May be harmful if swallowed.
    Chemical Name or Material Ruthenium(IV) oxide
    Grade Electronic Grade

    H271-H303: May cause fire or explosion; strong oxidizer. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit