Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(IV) oxide hydrate, > 54% Ru, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
32740-79-7
O2Ru·xH2O
133.07
MFCD00149846
32740-79-7
O2Ru·xH2O
133.07
MFCD00149846
Linear Formula RuO2·xH2O
Fieser 01,984; 14,272; 04,108; 05,161
Solubility Information Solubility in water: insoluble
Formula Weight 133.07
Physical Form Fine Powder
Percent Purity >54% Ru
Packaging Glass bottle
Solubility Solubility in water: insoluble.
Color Black
Quantity 25g
Chemical Name or Material Ruthenium(IV) oxide hydrate, >54% Ru
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning