Alfa Aesar™ Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%

Manufacturer: Alfa Aesar™ 4340306

Quantity 5g

 View more versions of this product

Catalog No. AA4340306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32740-79-7
    Molecular Formula RuO2·xH2O
    Formula Weight 133.07 (Anhydrous)
    MDL Number MFCD00149846
    UN Number UN1479
    Chemical Name or Material Ruthenium (IV) oxide hydrate
    Grade Premion™

    Specifications

    Formula Weight 133.07 (Anhydrous)
    Melting Point 1,200°C (subl.)
    Health Hazard 3 GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 32740-79-7
    Health Hazard 1 Danger
    Molecular Formula RuO2·xH2O
    Concentration Ru 54 to 60%
    MDL Number MFCD00149846
    Quantity 5g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Physical Form Powder
    UN Number UN1479
    Color Black
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H271
    May cause fire or explosion
    strong oxidizer.
    Chemical Name or Material Ruthenium (IV) oxide hydrate
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H271
    May cause fire or explosion
    strong oxidizer.

    GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit