Alfa Aesar™ Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%

Manufacturer: Alfa Aesar™ 4340314

Quantity 25g

 View more versions of this product

Catalog No. AA4340314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32740-79-7
    Molecular Formula RuO2·xH2O
    Formula Weight 133.07 (Anhydrous)
    MDL Number MFCD00149846
    UN Number UN1479
    Chemical Name or Material Ruthenium (IV) oxide hydrate
    Grade Premion™

    Specifications

    Formula Weight 133.07 (Anhydrous)
    Melting Point 1,200°C (subl.)
    Health Hazard 3 GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1479
    Concentration Ru 54 to 60%
    MDL Number MFCD00149846
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H271
    May cause fire or explosion
    strong oxidizer.
    TSCA TSCA
    Physical Form Powder
    Molecular Formula RuO2·xH2O
    CAS 32740-79-7
    Color Black
    Quantity 25g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Chemical Name or Material Ruthenium (IV) oxide hydrate
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H271
    May cause fire or explosion
    strong oxidizer.

    GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit