Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(IV) oxide hydrate, Ru 54% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1180306

Quantity 5g

 View more versions of this product

Catalog No. AA1180306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

32740-79-7
Ru 54% min.
133.07 (Anhydrous)
Powder
O2Ru·xH2O
Ruthenium(IV) oxide hydrate

Specifications

32740-79-7
Ru 54% min.
Black powder
Odorless
5g
Powder
O2Ru·xH2O
133.07 (Anhydrous)
Black
Ruthenium(IV) oxide hydrate

Safety and Handling

SDS & Safety Documents

H271: May cause fire or explosion; strong oxidizer.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 234-840-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit