Ruthenium plasma standard solution, Ru 10,000µg/mL, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 35773AE

Quantity 100mL

 View more versions of this product

Catalog No. AA35773AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula RuCl3 in 20% HCl
    MDL Number MFCD00011207
    Chemical Name or Material Ruthenium, Ru 10, 000μg/mL

    Specifications

    Quantity 100mL
    DOT Information DOT Class: 8; Packing Group: II
    EINECS Number 231-595-7
    Physical Form Liquid
    Molecular Formula RuCl3 in 20% HCl
    MDL Number MFCD00011207
    Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
    Appearance Liquid
    Chemical Name or Material Ruthenium, Ru 10, 000μg/mL

    H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit