Ruthenium powder, 99.>99% (metals basis excluding Ca), Alfa Aesar™, Premion™

Manufacturer: Alfa Aesar 4487914

Quantity 25g

 View more versions of this product

Catalog No. AA4487914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-18-8
    Physical Form Powder
    Assay Percent Range >99.99% (Metals basis excluding Ca)
    MDL Number MFCD00011207
    Chemical Name or Material Ruthenium

    Specifications

    Quantity 25g
    Color Gray
    DOT Information DOT Class: 4.1, Packaging Group: III
    Density 12.45g/cm3
    EINECS Number 231-127-1
    Physical Form Powder
    Assay Percent Range >99.99% (Metals basis excluding Ca)
    MDL Number MFCD00011207
    CAS 7440-18-8
    Odor Odorless
    Appearance gray powder
    Health Hazard 1 H228: Flammable solid.
    Chemical Name or Material Ruthenium

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit