Ruthenium Powder, Alfa Aesar™

Manufacturer: Alfa Aesar 1175706

Quantity 5g

 View more versions of this product

Catalog No. AA1175706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-18-8
    Physical Form Powder
    MDL Number MFCD00011207
    Chemical Name or Material Ruthenium

    Specifications

    Quantity 5g
    Color Black
    DOT Information DOT Class: 4.2, Packaging Group: III
    EINECS Number 231-127-1
    Physical Form Powder
    MDL Number MFCD00011207
    CAS 7440-18-8
    Odor Odorless
    Health Hazard 1 H228-H252-H315-H319-H335: Flammable solid. Self-heating in large quantities; may catch fire. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Appearance Black powder
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
    Chemical Name or Material Ruthenium

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

    H228-H252-H315-H319-H335: Flammable solid. Self-heating in large quantities; may catch fire. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit