Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ruv B Antibody (RuvB 4G5/1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250279H
Description
Ruv B Monoclonal antibody specifically detects Ruv B in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Ruv B | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Ruv B protein from E. Coli | |
0.1 mL | |
DNA Repair, Epigenetics, Virology Bacteria and Parasites | |
E. coli | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
RuvB 4G5/1 | |
Western Blot, ELISA, Immunohistochemistry | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction