Learn More
RuvB Mouse anti-Bacteria, Janelia Fluor 549, Clone: RuvB 4G5/1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250279JF549
Catalog No. NBP50279549
Add to cart
Description
Description
RuvB Monoclonal antibody specifically detects RuvB in Bacteria samples. It is validated for Immunohistochemistry.
Specifications
Specifications
RuvB
RuvB 4G5/1
Immunohistochemistry
Purified
Ruv B protein from E. Coli
Protein G purified
Store at 4°C in the dark.
Monoclonal
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
0.1mL
Primary
Bacteria