Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL1 Mouse anti-Human, Clone: 1D6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259456100UL

 View more versions of this product

Catalog No. NBP259456X


Add to cart

Description

Description

RUVBL1 Monoclonal antibody specifically detects RUVBL1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

RUVBL1
1D6
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
49 kDa TATA box-binding protein-interacting protein, 54 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,49 kDa TBP-interacting protein, ECP54, INO80 complex subunit H, INO80HTIP49A, NMP 238, NMP238ECP-54, Nuclear matrix protein 238, PONTIN, Pontin 52, Pontin52, RuvB (E coli homolog)-like 1, ruvB-like 1, RuvB-like 1 (E. coli), RVB1, TATA binding protein interacting protein 49 kDa, TIH1, TIP49a, TIP49TAP54-alpha, TIP60-associated protein 54-alpha
Mouse
IgG2a Kappa
100 ul
Core ESC Like Genes, Stem Cell Markers, Wnt Signaling Pathway
Primary
8607
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1:10-1:1058, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Purified
RUVBL1
Recombinant human RuVBL1 (1-456aa) purified from E. coli
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only