Learn More
RUVBL2 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1D9, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504285
Catalog No. 50-167-5006
Add to cart
Description
Description
Possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (5 ft. to 3 ft.) activity. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL2 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1-CTNNB1 complex. May also inhibit the transcriptional activity of ATF2.
Specifications
Specifications
RUVBL2
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9WTM5, Q9Y230
CGI-46, ECP-51, ECP51, INO80J, REPTIN, RVB2, TAP54-beta, TIH2, TIP48, TIP49B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10856, 20174, 292907, 476418
Immunohistochemistry (Paraffin), Western Blot
OTI1D9
Unconjugated
RUVBL2
Liquid
Ruvbl2
Human recombit protein fragment corresponding to amino acids 113-370 of human RUVBL2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat