Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ RVZ22SE Series - Self Extinguishing Safety Vest

Pyramex™ RVZ22SE Series - Self Extinguishing Safety Vest with ANSI/ISEA 107-2015 safety compliance

Manufacturer:  Pyramex™ RVZ2210SEM

 View more versions of this product

Catalog No. 19-555-516



Add to cart
This item is not returnable. View return policy

Description

Description

  • RVZ22SE Series vest is not flame resistant but it will self-extinguish
  • One 3.5 in. contrasting material stripe applied around vest horizontally 2.5 in. from bottom, with 2 in. silver reflective strip centered inside contrasting stripe
  • Two 3.5 in. contrasting material stripes with 2 in. silver reflective strip centered over contrasting strip to run vertically from bottom stripe, over shoulders to the stripe on the back side of the vest
  • Lightweight polyester mesh material
  • Zipper front closure
  • Chest pocket (5.5 x 6.5 in.) with hook and loop closure
  • Deep and wide inside pocket (7.5 x 7.5 in)
  • Individually packaged
  • 50 vests per case
Specifications

Specifications

26 in.
25 in.
ANSI/ISEA 107-2015, Type R - Class 2
China Brand Silver
Medium
2
Polyester
Zipper
Hi-Vis Lime
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit