Learn More
RWDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170361AP
Catalog No. 50-173-1341
Add to cart
Description
Description
The Formin-like protein 1 (FRL1) gene encodes a formin-related protein which has been implicated in morphogenesis, cyokinesis, and cell polarity. Formins are a conserved class of proteins expressed in all eukaryotes and have one DAD (diaphanous autoregulatory domain), one FH2 (formin homology 2) domain and one GBD/FH3 (Rho GTPase-binding / formin homology 3) domain. FRL1 is located in the cytoplasm and is highly expressed in the spleen, lymph node and bone marrow cells. FRL1 possibly has a role in the control of cell motility, survival of macrophages and cytoskeletal organization.
Specifications
Specifications
RWDD1
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99ND9, Q9CQK7, Q9H446
CGI 24, DFRP2, PTD013, RWD domain containing 1, RWDD1
Rabbit
IgG
150 μL
-20°C
Primary
259218, 51389, 66521
Immunoprecipitation, Western Blot
Unconjugated
RWDD1
Liquid
RWDD1
RWDD1 Fusion Protein Ag10525
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat