Learn More
Aviva Systems Biology Corp RWDD4A Antibody - middle region (ARP53106_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53106_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RWDD4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RWDD4A. Alternate names: FAM28A, MGC10198, RWDD4AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60923