Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RWJ 21757, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
121288-39-9
C13H17N5O6
339.308
VDCRFBBZFHHYGT-FCKMSMMTSA-N
7, 8-Dihydro-8-oxo-7-(2-propenyl)guanosine
122173106
2-amino-9-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7-prop-2-enyl-3H-purine-6,8-dione
C=CCN1C2=C(NC(=NC2=O)N)N(C1=O)C3C(C(C(O3)CO)O)O
This item is not returnable. View return policy
121288-39-9
C13H17N5O6
339.308
VDCRFBBZFHHYGT-FCKMSMMTSA-N
7, 8-Dihydro-8-oxo-7-(2-propenyl)guanosine
122173106
2-amino-9-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7-prop-2-enyl-3H-purine-6,8-dione
C=CCN1C2=C(NC(=NC2=O)N)N(C1=O)C3C(C(C(O3)CO)O)O

Toll-like receptor 7 (TLR7) agonist

Toll-like receptor 7 (TLR7) agonist; induces immune cell activation and increases cytokine production. Displays antitumor and antiviral activity in various animal models.
Quantity 50mg
Formula Weight 339.3
Percent Purity >98%
Chemical Name or Material RWJ 21757

Recommended Storage : Desiccate at +4°C