Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RWJ 21757, Tocris Bioscience™

Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Toll-like receptor 7 (TLR7) agonist

Toll-like receptor 7 (TLR7) agonist; induces immune cell activation and increases cytokine production. Displays antitumor and antiviral activity in various animal models.
CAS 121288-39-9
Molecular Formula C13H17N5O6
Quantity 10 mg
Synonym 7, 8-Dihydro-8-oxo-7-(2-propenyl)guanosine
InChI Key VDCRFBBZFHHYGT-FCKMSMMTSA-N
SMILES C=CCN1C2=C(NC(=NC2=O)N)N(C1=O)C3C(C(C(O3)CO)O)O
IUPAC Name 2-amino-9-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7-prop-2-enyl-3H-purine-6,8-dione
Molecular Weight (g/mol) 339.308
PubChem CID 122173106
Formula Weight 339.3
Percent Purity >98%
Chemical Name or Material RWJ 21757
Show More Show Less

Recommended Storage : Desiccate at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit