Learn More
Selleck Chemical LLCSupplier Diversity Partner RXDX-106 (CEP-40783) DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85705MG
RXDX-106 (CEP-40783) is an orally-available, potent and selective TAM(TYRO3, AXL, MER)/MET inhibitor displaying low nanomolar biochemical activity and slow (T1/2 >120 min) inhibitor off-rate in peptide phosphorylation assays and in vitro kinase binding assays, respectively.
Catalog No. 50-194-8114