Learn More
RXFP4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205501AP
Catalog No. 50-173-2955
Add to cart
Description
Description
GPR100 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) and acts as a high affinity receptor for INSL5. Also acts as receptor for RLN3/relaxin-3, as well as bradykinin and kallidin. Binding of the ligand inhibits cAMP accumulation.
Specifications
Specifications
RXFP4
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8TDU9
G protein coupled receptor 100, GPCR142, GPR100, Relaxin 3 receptor 2, RLN3 receptor 2, RLN3R2, RXFP4, RXFPR4
Rabbit
IgG
150 μL
-20°C
Primary
339403
Immunohistochemistry (Paraffin)
Unconjugated
RXFP4
Liquid
RXFP4
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human