Learn More
Restek™ Rxi™-17 Capillary Columns
General-purpose columns for pesticides, herbicides, rosin acids, phthalate esters, triglycerides, and sterols
$398.00 - $909.00
Specifications
For Use With (Application) | Pesticides, Herbicides, Rosin Acids, Phthalate Esters, Triglycerides, Sterols |
---|---|
Stationary Phase | Crossbond Diphenyl Dimethyl Polysiloxane |
Temperature Range | 40°C to 320°C |
Packing Material | Rxi™-17 |
Polarity | Midpolar |
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
06-611-004
![]() |
Restek™
13502 |
0.18 mm | 0.18 μm | 20 m |
Each for $585.00
|
|
06-611-005
![]() |
Restek™
13520 |
0.25 mm | 0.25 μm | 15 m |
Each for $398.00
|
|
06-611-006
![]() |
Restek™
13523 |
0.25 mm | 0.25 μm | 30 m |
Each for $647.00
|
|
06-611-009
![]() |
Restek™
13538 |
0.25 mm | 0.5 μm | 30 m |
Each for $647.00
|
|
06-611-013
![]() |
Restek™
13553 |
0.25 mm | 1 μm | 30 m |
Each for $647.00
|
|
06-611-007
![]() |
Restek™
13524 |
0.32 mm | 0.25 μm | 30 m |
Each for $813.00
|
|
06-611-010
![]() |
Restek™
13539 |
0.32 mm | 0.5 μm | 30 m |
Each for $787.00
|
|
06-611-014
![]() |
Restek™
13554 |
0.32 mm | 1 μm | 30 m |
Each for $787.00
|
|
06-611-008
![]() |
Restek™
13525 |
0.53 mm | 0.25 μm | 30 m |
Each for $881.00
|
|
06-611-011
![]() |
Restek™
13540 |
0.53 mm | 0.5 μm | 30 m |
Each for $909.00
|
|
06-611-012
![]() |
Restek™
13552 |
0.53 mm | 1 μm | 15 m |
Each for $450.00
|
|
06-611-015
![]() |
Restek™
13555 |
0.53 mm | 1 μm | 30 m |
Each for $881.00
|
|
06-611-016
![]() |
Restek™
13570 |
0.53 mm | 1.5 μm | 30 m |
Each for $881.00
|
|
Description
- Ultra-low bleed
- Exceptional inertness
- Reproducible performance, column after column
- Temperature range: 40°C to 320°C
Specifications
Pesticides, Herbicides, Rosin Acids, Phthalate Esters, Triglycerides, Sterols | |
40°C to 320°C | |
Midpolar | |
1 |
Crossbond Diphenyl Dimethyl Polysiloxane | |
Rxi™-17 | |
Fused Silica |