Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Rxi™-1HT Capillary Columns

Columns processed for high-temperature applications, such as high molecular weight hydrocarbons
$501.00 - $886.00
Specifications
For Use With (Application) | High Molecular Weight Hydrocarbons, High-Temperature Applications |
---|---|
Stationary Phase | 100% Dimethyl Polysiloxane |
Temperature Range | -60°C to +400°C |
Packing Material | Rxi™-1 HT |
Polarity | Nonpolar |
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
06-611-060
![]() |
Restek™
13950 |
0.25 mm | 0.1 μm | 15 m |
Each for $501.00
|
|
06-611-061
![]() |
Restek™
13951 |
0.25 mm | 0.1 μm | 30 m |
Each for $886.00
|
|
06-611-062
![]() |
Restek™
13952 |
0.25 mm | 0.25 μm | 30 m |
Each for $884.00
|
|
06-611-063
![]() |
Restek™
13953 |
0.32 mm | 0.1 μm | 15 m |
Each for $614.00
|
|
06-611-064
![]() |
Restek™
13954 |
0.32 mm | 0.1 μm | 30 m |
Each for $836.00
|
|
06-611-065
![]() |
Restek™
13955 |
0.32 mm | 0.25 μm | 30 m |
Each for $836.00
|
|
Description
- Ultra-low bleed
- Exceptional inertness
- Reproducible performance, column after column
Specifications
High Molecular Weight Hydrocarbons, High-Temperature Applications | |
-60°C to +400°C | |
Nonpolar | |
1 Column |
100% Dimethyl Polysiloxane | |
Rxi™-1 HT | |
Fused Silica |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title