Learn More
Restek™ Rxi-65TG Columns (Fused Silica)
High-polarity crossbond phase
Supplier: Restek™ 17109
Description
- Ideal for analysis of triglycerides in edible oils
- Thermal stability up to 370 °C ensures consistent results and longer column lifetimes
- Separate and quantify critical triglycerides (acylglycerols) without interference from column bleed
- Observe even underivatized mono- and diglycerides
- Novel manufacturing techniques ensure consistent column-to-column quality
Note:
- Rxi-65TG column is recommended for analyzing mono-, di-, or triglycerides in edible oils
- If analyzing triglycerides in biodiesel fuels, use either Rtx-Biodiesel TG or MXT-Biodiesel TG columns
Specifications
0.32 mm | |
Analysis of triglycerides in edible oils, separate and quantify critical triglycerides, observe undervatized mono- and diglycerides | |
0.1 μm | |
Silica |
Rxi-65 TG column | |
40 to 370°C | |
30 m |