Learn More
Restek™ Rxi™ Guard/Retention Gap Columns
SDP
Extend column lifetime
Supplier:  Restek™ 10073600
 View more versions of this product
Catalog No. 06-610-583
Add to cart
Description
Description
  • Extends column lifetime
  • Excellent inertness achieves lower detection limits for active compounds
  • Sharper chromatographic peaks by utilizing retention gap technology
  • Maximum temperature: 360°C
Specifications
Specifications
Rxi™ Columns
0.53 mm
Fused Silica
Guard/Retention Gap Columns
10 m
6/Pk.