Learn More
RXRA Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ARC0468, Invitrogen™
Rabbit Monoclonal Antibody
Manufacturer:  Invitrogen MA535265
Catalog No. PIMA535265
Add to cart
Description
Description
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
Specifications
Specifications
RXRA
ARC0468
PBS with 50% glycerol and 0.2% sodium azide; pH 7.3
P19793, P28700, Q05343
9530071D11Rik; NR2B1; Nuclear receptor subfamily 2 group B member 1; Retinoic acid receptor RXR-alpha; retinoid X receptor; Retinoid X receptor alpha; RXRA; RXRalpha1
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
20181, 25271, 6256
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rxra
Liquid
Rxra
A synthesized peptide derived from human Retinoid X Receptor alpha
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Rat