Learn More
Sklar InstrumentsSupplier Diversity Partner TC Ryder Needle Holder, Sklar, 5 in., 2mm, Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218019
TC Ryder Needle Holder, Sklar, 5 in., 2mm, Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's TC Ryder Needle Holder is a ratcheted, finger ring instrument that features narrow jaws and is commonly used with very small suture needles in cardiovascular, plastic, and neurosurgical procedures. Tungsten carbide cross-serrated tips firmly grip the small needle and assist with keeping it from twisting during suturing. This product is straight with 2mm wide, cross-serrated jaws, and an overall length of 5 inches.
Catalog No. 50-143-7707