Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1,1'-Bi(2-naphthyl diacetate), 98%, Acros Organics

Click to view available options
Quantity:
5g
69677-98-1
C24H22O4
374.436
MFCD00691542
DJUHRILTVUZDIO-UHFFFAOYSA-N
2,2'-diacetoxy-1,1'-binaphthyl,1-2-acetyloxynaphthyl-2-naphthyl acetate,1-2-acetoxy-1-naphthyl-2-naphthyl acetate,1,1'-bi 2-naphthol diacetate,r--1,1'-bi-2-naphthol deacetate,1,1'-binaphthalene-2,2'-diyl diacetate,r---2,2'-diacetoxy-1,1'-binaphthyl,s---2,2'-diacetoxy-1,1'-binaphthyl,r-1,1'-binaphthalene-2,2'-diyl diacetate,s-1,1'-binaphthalene-2,2'-diyl diacetate
24820722
acetic acid;1-naphthalen-1-ylnaphthalene
CC(=O)O.CC(=O)O.C1=CC=C2C(=C1)C=CC=C2C3=CC=CC4=CC=CC=C43
69677-98-1
C24H22O4
374.436
MFCD00691542
DJUHRILTVUZDIO-UHFFFAOYSA-N
2,2'-diacetoxy-1,1'-binaphthyl,1-2-acetyloxynaphthyl-2-naphthyl acetate,1-2-acetoxy-1-naphthyl-2-naphthyl acetate,1,1'-bi 2-naphthol diacetate,r--1,1'-bi-2-naphthol deacetate,1,1'-binaphthalene-2,2'-diyl diacetate,r---2,2'-diacetoxy-1,1'-binaphthyl,s---2,2'-diacetoxy-1,1'-binaphthyl,r-1,1'-binaphthalene-2,2'-diyl diacetate,s-1,1'-binaphthalene-2,2'-diyl diacetate
24820722
acetic acid;1-naphthalen-1-ylnaphthalene
CC(=O)O.CC(=O)O.C1=CC=C2C(=C1)C=CC=C2C3=CC=CC4=CC=CC=C43
Quantity 5g
Color White
Specific Rotation +15° to +19° (20°C, 589nm) (c=1, THF)
Infrared Spectrum Authentic
Melting Point 111°C to 116°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 370.4
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material (S)-(+)-1, 1'-Bi(2-naphthyl diacetate)
Show More Show Less