Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulfonate), 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
128544-05-8
C22H12F6O6S2
550.442
MFCD00274615
OYJLCOSEYYZULE-UHFFFAOYSA-N
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate,s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate,r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate,r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate,s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate,r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate,s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate,1,1'-bi-2-naphthol bis trifluoromethanesulfonate,s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate,r---binol-tf2
389883
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
128544-05-8
C22H12F6O6S2
550.442
MFCD00274615
OYJLCOSEYYZULE-UHFFFAOYSA-N
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate,s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate,r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate,r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate,s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate,r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate,s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate,1,1'-bi-2-naphthol bis trifluoromethanesulfonate,s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate,r---binol-tf2
389883
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
Infrared Spectrum Authentic
Quantity 5g
Color White to Yellow
Melting Point 80°C to 85°C
Specific Rotation +145° to +149° (20°C, 589nm) (c=1, CHCl3)
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CF3SO3C10H6-)2
Formula Weight 550.44
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material (S)-(+)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA