Learn More
Alfa Aesar™ (S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride, 95%
Manufacturer:  Alfa Aesar™ H52111MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH52111MD
  Check Availability
Add to cart
Specifications
Description
Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications
Chemical Identifiers
270082-22-9
227.69
H-Tqa-OH·HCl
C11H13NO2·HCl
MFCD01861030
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride
Specifications
250mg
C11H13NO2·HCl
MFCD01861030
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride
270082-22-9
227.69
H-Tqa-OH·HCl
95%