Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol, 95%

Click to view available options
Quantity:
250mg
1g
5g
877397-65-4
C8H7Cl2FO
209.041
MFCD09863793
JAOYKRSASYNDGH-BYPYZUCNSA-N
s-1-2,6-dichloro-3-fluorophenyl ethanol,1s-1-2,6-dichloro-3-fluorophenyl ethanol,1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol,s-1-2,6-dichloro-3-fluorophenyl-ethanol,pubchem17375,1s-1-2,6-dichloro-3-fluoro-phenyl ethanol,ksc497a2n,s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol,s-1-2',6'-dichloro-3'-fluorophenyl-ethanol,1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol 30:70 r:s
11557536
(1S)-1-(2,6-dichloro-3-fluorophenyl)ethanol
CC(C1=C(C=CC(=C1Cl)F)Cl)O
877397-65-4
C8H7Cl2FO
209.041
MFCD09863793
JAOYKRSASYNDGH-BYPYZUCNSA-N
s-1-2,6-dichloro-3-fluorophenyl ethanol,1s-1-2,6-dichloro-3-fluorophenyl ethanol,1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol,s-1-2,6-dichloro-3-fluorophenyl-ethanol,pubchem17375,1s-1-2,6-dichloro-3-fluoro-phenyl ethanol,ksc497a2n,s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol,s-1-2',6'-dichloro-3'-fluorophenyl-ethanol,1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol 30:70 r:s
11557536
(1S)-1-(2,6-dichloro-3-fluorophenyl)ethanol
CC(C1=C(C=CC(=C1Cl)F)Cl)O
Quantity 5g
Color White
Formula Weight 209.04
Physical Form Solid
Percent Purity 95%
Chemical Name or Material (S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol