Alfa Aesar™ (S)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%

Manufacturer: Alfa Aesar™ H2694806

Quantity 5g

 View more versions of this product

Catalog No. AAH2694806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68285-24-5
    Molecular Formula C9H13NO
    Formula Weight 151.21
    MDL Number MFCD00671658
    UN Number UN2735
    Synonym (S)-o-Methoxy-alpha-methylbenzylamine
    Chemical Name or Material (S)-1-(2-Methoxyphenyl)ethylamine

    Specifications

    Formula Weight 151.21
    UN Number UN2735
    Molecular Formula C9H13NO
    MDL Number MFCD00671658
    CAS 68285-24-5
    Synonym (S)-o-Methoxy-alpha-methylbenzylamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 118°C (17mmHg)
    Quantity 5g
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (S)-1-(2-Methoxyphenyl)ethylamine
    Assay 99%, ee ≥98%

    Description

    Applications
    (S)-1-(2-Methoxyphenyl)ethylamine play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

    Solubility
    Soluble in water(22 g/L).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

    GHS H Statement
    H301-H311-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit